Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Aminoquinoline | 580-15-4 | 144.17 | 10 G
SDP

Catalog No. 5000460385
Encompass_Preferred

6-Aminoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Appears as a solid
  • Ranges in color from light yellow to brown
  • Soluble in DMSO at concentrations of 200 mg/mL or greater
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-60385 Supplier Medchemexpress LLC Supplier No. HYW00740410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Aminoquinoline is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Appears as a solid
  • Ranges in color from light yellow to brown
  • Soluble in DMSO at concentrations of 200 mg/mL or greater
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.