Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Aminoquinoline | 580-15-4 | MFCD00006803 | 144.17 | 25 G
SDP

Catalog No. 5000449977
Encompass_Preferred

6-Aminoquinoline is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only.

  • Biochemical reagent for life science research
  • Solid appearance, light yellow to brown color
  • Purity of 99.79%
  • Molecular weight of 144.17
  • Store at 4°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)

Catalog No. 50-004-49977 Supplier Medchemexpress LLC Supplier No. 25GHYW007404
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Aminoquinoline is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only.

  • Biochemical reagent for life science research
  • Solid appearance, light yellow to brown color
  • Purity of 99.79%
  • Molecular weight of 144.17
  • Store at 4°C, protect from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.