Learn More
Gold Biotechnology Inc 6-Aminoquinolyl-N-hydroxysuccinimidyl carbamate 5 mg
SDP
Supplier:  Gold Biotechnology Inc A6755
Encompass
6-Aminoquinolyl-N-hydroxysuccinimidyl carbamate is used to detect proteins and amino acids. It reacts with amino acids forming stable unsymmetrical urea derivatives. It can be used with HPLC analysis.
Catalog No. NC1791914