Learn More
Medchemexpress LLC 6-aminoquinolyl-N-hydroxysuccinimidyl carbamate | 148757-94-2 | MFCD01940654 | 95.0% | 285.25 g/mol | C14H11N3O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11769525MG
Encompass_Preferred
AQC (6-aminoquinolyl-N-hydroxysuccinimidyl carbamate) is a fluorogenic derivatization reagent used to label primary and secondary amines for HPLC and fluorescence detection. It forms stable fluorescent urea derivatives that enable sensitive amino acid and protein analysis. Supplied as a solid powder with a specified purity for analytical use.
  • Reacts rapidly with primary and secondary amines to form fluorescent derivatives.
  • Enhances sensitivity and selectivity in HPLC fluorescence detection.
  • Produces stable derivatives suitable for quantitation and sequencing workflows.
  • Available in small vial sizes for analytical and method-development work.
  • Supplied as a solid with defined purity to support reproducible results.
Catalog No. 50-002-28346