Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminoquinoxaline 98.0+%, TCI America™
SDP

Catalog No. A24905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24905G 5 g
A24901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24905G Supplier TCI America Supplier No. A24905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6298-37-9
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00462821
InChI Key MSGRFBKVMUKEGZ-UHFFFAOYSA-N
Synonym 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino
PubChem CID 237859
IUPAC Name quinoxalin-6-amine
SMILES NC1=CC=C2N=CC=NC2=C1

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 145.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminoquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.