Learn More
AdipoGen 6-Aminoseleno-D-luciferin
SDP
Catalog No. 502561756
Encompass_Preferred
Chemical. CAS 1375964-28-5. Formula C11H9N3O2SSe. MW 326.23. Synthetic. Firefly luciferase substrate. Selenium analog of firefly L-luciferin with red-shifted bioluminescence emission for use in in vivo bioluminescence imaging. Bioluminescence spectra max. 600nm. This selenium analog exhibits a similar substrate affinity to wild type D-luciferin, though with slightly lower quantum yield.
Catalog No. 50-256-1756 Supplier AdipoGen Supplier No. CDXA0307M005
Add to Cart
Add to Cart
Chemical. CAS 1375964-28-5. Formula C11H9N3O2SSe. MW 326.23. Synthetic. Firefly luciferase substrate. Selenium analog of firefly L-luciferin with red-shifted bioluminescence emission for use in in vivo bioluminescence imaging. Bioluminescence spectra max. 600nm. This selenium analog exhibits a similar substrate affinity to wild type D-luciferin, though with slightly lower quantum yield.