Learn More
Sigma Aldrich Fine Chemicals Biosciences 6 Amyl alpha pyrone 96 fG1KG
Catalog No. 502914358
Encompass
6-Amyl -pyrone occurs naturally in peaches. It is also the main volatile organic compound produced by Trichoderma species.
Catalog No. 50-291-4358 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3696081KGK
Add to Cart
Add to Cart
Request a Quote
6-Amyl -pyrone occurs naturally in peaches. It is also the main volatile organic compound produced by Trichoderma species.