Learn More
Cayman Chemical 6-APDB (hydrochloride), Purity: ≥95%, CAS: 1281872-58-9, 10 mg
SDP
Catalog No. 502951504
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; substitutes for (S)-(+)-MBDB and MMAI in two-lever drug discrimination tests in rats; intended for research and forensic applications
Catalog No. 50-295-1504 Supplier Cayman Chemical Supplier No. 1114610 MG
An analytical reference standard categorized as a benzofuran; substitutes for (S)-(+)-MBDB and MMAI in two-lever drug discrimination tests in rats; intended for research and forensic applications