Learn More
Medchemexpress LLC 6-Azathymine | 932-53-6 | 99.9% | 127.10 | 250 MG
SDP
Catalog No. 5000352282
Encompass_Preferred
6-Azathymine, a 6-nitrogen analog of thymine, is a potent D-3-aminoisobutyrate-pyruvate aminotransferase inhibitor. It inhibits the biosynthesis of DNA and possesses antibacterial and antiviral activities.
  • Acts as a competitive antagonist against the growth of microorganisms.
  • Inhibits the biosynthesis of DNA.
  • Possesses antibacterial and antiviral activities.
  • Undergoes urinary elimination of both free azathymine and its metabolites in vivo.
  • Radioactivity from azathymine-5-14C is found in all investigated tissues.
Catalog No. 50-003-52282 Supplier Medchemexpress LLC Supplier No. 250MGHY136559
6-Azathymine, a 6-nitrogen analog of thymine, is a potent D-3-aminoisobutyrate-pyruvate aminotransferase inhibitor. It inhibits the biosynthesis of DNA and possesses antibacterial and antiviral activities.
  • Acts as a competitive antagonist against the growth of microorganisms.
  • Inhibits the biosynthesis of DNA.
  • Possesses antibacterial and antiviral activities.
  • Undergoes urinary elimination of both free azathymine and its metabolites in vivo.
  • Radioactivity from azathymine-5-14C is found in all investigated tissues.