Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-azauridine 5'-triphosphate ammonium | 98.0% | 502.16 | C8H17N4O15P3 | 10 MG
SDP

Catalog No. 5000286636
Encompass_Preferred

6-Azauridine 5′-Triphosphate ammonium is a nucleotide analog of uridine triphosphate supplied as the ammonium salt for biochemical and molecular biology research. The material is provided as a white to off-white solid in a 10 mg presentation with 98.0% purity and is used to study RNA synthesis, transcriptional regulation, and nucleotide metabolism. Store under nitrogen and away from moisture; in solvent, store at -80°C for up to 6 months or -20°C for 1 month.

  • Nucleotide analog of uridine triphosphate for biochemical assays.
  • Supplied as a white to off-white solid powder.
  • High purity: 98.0%.
  • Compact 10 mg presentation for small-scale experiments.
  • Molecular weight 502.16 and formula C8H17N4O15P3.
  • Storage instructions: store under nitrogen and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-86636 Supplier Medchemexpress LLC Supplier No. HY154909A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Azauridine 5′-Triphosphate ammonium is a nucleotide analog of uridine triphosphate supplied as the ammonium salt for biochemical and molecular biology research. The material is provided as a white to off-white solid in a 10 mg presentation with 98.0% purity and is used to study RNA synthesis, transcriptional regulation, and nucleotide metabolism. Store under nitrogen and away from moisture; in solvent, store at -80°C for up to 6 months or -20°C for 1 month.

  • Nucleotide analog of uridine triphosphate for biochemical assays.
  • Supplied as a white to off-white solid powder.
  • High purity: 98.0%.
  • Compact 10 mg presentation for small-scale experiments.
  • Molecular weight 502.16 and formula C8H17N4O15P3.
  • Storage instructions: store under nitrogen and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.