Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Azauridine triphosphate ammonium | C8H17N4O15P3 | 1 MG
SDP

Catalog No. 5000376079
Encompass_Preferred

6-Azauridine triphosphate ammonium is a nucleotide analog similar to uridine triphosphate. It is used to study the mechanism of RNA synthesis and transcription regulation.

  • Purity of 95.0%
  • Appears as a solid
  • Color is white to off-white
  • Soluble in H2O at 250 mg/mL (497.85 mM; requires ultrasonic)

Catalog No. 50-003-76079 Supplier Medchemexpress LLC Supplier No. HY154909A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Azauridine triphosphate ammonium is a nucleotide analog similar to uridine triphosphate. It is used to study the mechanism of RNA synthesis and transcription regulation.

  • Purity of 95.0%
  • Appears as a solid
  • Color is white to off-white
  • Soluble in H2O at 250 mg/mL (497.85 mM; requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.