Learn More
Medchemexpress LLC 6-Azauridine triphosphate ammonium | C8H17N4O15P3 | 5 MG
SDP
Catalog No. 5000376080
Encompass_Preferred
6-Azauridine triphosphate ammonium is a nucleotide analog that resembles uridine triphosphate. It is utilized in research to investigate the mechanisms of RNA synthesis and the regulation of transcription.
  • Used to study RNA synthesis
  • Used to study transcription regulation
Catalog No. 50-003-76080 Supplier Medchemexpress LLC Supplier No. HY154909A5MG
6-Azauridine triphosphate ammonium is a nucleotide analog that resembles uridine triphosphate. It is utilized in research to investigate the mechanisms of RNA synthesis and the regulation of transcription.
  • Used to study RNA synthesis
  • Used to study transcription regulation