Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-azido-1-hexanamine | 349553-73-7 | MFCD07369845 | 98.7% | 142.20 g/mol | C6H14N4 | 500 MG
SDP

Catalog No. 5000203891
Encompass_Preferred

6-azido-1-hexanamine is an azide-functionalized hexylamine reagent used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and for click chemistry conjugation. It provides an azide handle for azide-alkyne cycloaddition reactions and is supplied as a colorless to light yellow liquid.

  • CAS number 349553-73-7
  • Molecular formula C6H14N4
  • Molecular weight 142.20 g/mol
  • Purity 98.7%
  • Pack size 500 mg
  • Storage 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)
  • Applications ADC linker synthesis and click chemistry conjugation

Catalog No. 50-002-03891 Supplier Medchemexpress LLC Supplier No. HY138387500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-azido-1-hexanamine is an azide-functionalized hexylamine reagent used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and for click chemistry conjugation. It provides an azide handle for azide-alkyne cycloaddition reactions and is supplied as a colorless to light yellow liquid.

  • CAS number 349553-73-7
  • Molecular formula C6H14N4
  • Molecular weight 142.20 g/mol
  • Purity 98.7%
  • Pack size 500 mg
  • Storage 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month)
  • Applications ADC linker synthesis and click chemistry conjugation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.