Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-azido-1-hexanol | 146292-90-2 | MFCD22577171 | 98.0% | 143.19 | C6H13N3O | 50 MG
SDP

Catalog No. 5000202586
Encompass_Preferred

Azido-C6-OH is an azido-functionalized six-carbon linker used as a click-chemistry reagent and cleavable linker in antibody-drug conjugate and PROTAC synthesis. It provides an azide handle and a terminal hydroxyl for further derivatization.

  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Six-carbon spacer with terminal hydroxyl enables conjugation and derivatization.
  • High reported purity (98.0% by NMR) suitable for research workflows.
  • Molecular weight: 143.19, molecular formula: C6H13N3O.
  • Available in multiple pack sizes from manufacturers and distributors for synthetic applications.

Catalog No. 50-002-02586 Supplier Medchemexpress LLC Supplier No. HY13852150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-C6-OH is an azido-functionalized six-carbon linker used as a click-chemistry reagent and cleavable linker in antibody-drug conjugate and PROTAC synthesis. It provides an azide handle and a terminal hydroxyl for further derivatization.

  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Six-carbon spacer with terminal hydroxyl enables conjugation and derivatization.
  • High reported purity (98.0% by NMR) suitable for research workflows.
  • Molecular weight: 143.19, molecular formula: C6H13N3O.
  • Available in multiple pack sizes from manufacturers and distributors for synthetic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.