Learn More
Medchemexpress LLC 6-azido-6-deoxy-D-galactose | 66927-03-5 | MFCD03265529 | 98.2% | 205.17 | C6H11N3O5 | 50mg
SDP
Catalog No. 5000270089
Encompass_Preferred
6-Azido-6-deoxy-D-galactose 95% can be used as a substrate in enzymology to study the activity and specificity of galactosyltransferases and other glycosylation enzymes
Catalog No. 50-002-70089 Supplier Medchemexpress LLC Supplier No. HYW71813750MG
6-Azido-6-deoxy-D-galactose 95% can be used as a substrate in enzymology to study the activity and specificity of galactosyltransferases and other glycosylation enzymes