Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-azido-6-deoxy-D-glucose | 20847-05-6 | MFCD03265537 | 95.0% | 205.17 g·mol⁻¹ | C6H11N3O5 | 10 MG
SDP

Catalog No. 5000284643
Encompass_Preferred

6-Azido-6-deoxy-D-glucose is an azido-modified glucose analog used as a chemical reporter to visualize protein-specific glycosylation in living cells and as a reagent for biochemical assays. The azide at the 6-position enables bioorthogonal labeling (for example, click chemistry) of glycoconjugates in biological systems.

  • Purity: 95%
  • Molecular weight: 205.17 g·mol⁻¹
  • Chemical formula: C6H11N3O5
  • CAS number: 20847-05-6
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month)
  • Application: visualizing protein-specific glycosylation in living cells and biochemical assays

Catalog No. 50-002-84643 Supplier Medchemexpress LLC Supplier No. HYW67403910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Azido-6-deoxy-D-glucose is an azido-modified glucose analog used as a chemical reporter to visualize protein-specific glycosylation in living cells and as a reagent for biochemical assays. The azide at the 6-position enables bioorthogonal labeling (for example, click chemistry) of glycoconjugates in biological systems.

  • Purity: 95%
  • Molecular weight: 205.17 g·mol⁻¹
  • Chemical formula: C6H11N3O5
  • CAS number: 20847-05-6
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month)
  • Application: visualizing protein-specific glycosylation in living cells and biochemical assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.