Learn More
Medchemexpress LLC 6-Azido-6-deoxy-D-glucose | 20847-05-6 | MFCD03265537 | 95.0% | 205.17 g/mol | C6H11N3O5 | 50 MG
SDP
Catalog No. 5000284831
Encompass_Preferred
6-Azido-6-deoxy-D-glucose is an azide-functionalized glucose derivative used as a chemical reporter for metabolic labeling to visualize protein-specific glycosylation in living cells. Supplied as a powder with 95.0% purity, it is suitable for bioorthogonal click chemistry workflows and is stored under frozen conditions to preserve stability.
  • Azide-functionalized glucose for metabolic labeling of glycoconjugates.
  • Used to visualize protein-specific glycosylation in living cells.
  • Provided as a powder with 95.0% purity.
  • Molecular weight 205.17 g/mol; chemical formula C6H11N3O5.
  • Store powder at -20°C; in solvent store at -80°C for up to 6 months.
  • Suitable for azide-alkyne click chemistry tagging workflows.
Catalog No. 50-002-84831 Supplier Medchemexpress LLC Supplier No. HYW67403950MG
6-Azido-6-deoxy-D-glucose is an azide-functionalized glucose derivative used as a chemical reporter for metabolic labeling to visualize protein-specific glycosylation in living cells. Supplied as a powder with 95.0% purity, it is suitable for bioorthogonal click chemistry workflows and is stored under frozen conditions to preserve stability.
  • Azide-functionalized glucose for metabolic labeling of glycoconjugates.
  • Used to visualize protein-specific glycosylation in living cells.
  • Provided as a powder with 95.0% purity.
  • Molecular weight 205.17 g/mol; chemical formula C6H11N3O5.
  • Store powder at -20°C; in solvent store at -80°C for up to 6 months.
  • Suitable for azide-alkyne click chemistry tagging workflows.