Learn More
Medchemexpress LLC 6-azido-6-deoxy-d-glucose | 20847-05-6 | MFCD32206193 | 95.0% | 205.17 | C6H11N3O5 | 1 MG
SDP
Catalog No. 5000284946
Encompass_Preferred
6-Azido-6-deoxy-D-glucose (95%) is an azide-functionalized glucose analog used as a chemical reporter to visualize and study protein-specific glycosylation in living cells. The azido group at C-6 enables bioorthogonal labeling for downstream detection and imaging in cellular assays.
  • Azide functional group enables bioorthogonal labeling.
  • Suitable for live-cell visualization of protein glycosylation.
  • Supplied as a powder with recommended cold storage for stability.
  • High purity (95%) for consistent assay performance.
  • Defined molecular composition (C6H11N3O5, MW 205.17) for accurate experimental planning.
Catalog No. 50-002-84946 Supplier Medchemexpress LLC Supplier No. HYW6740391MG
6-Azido-6-deoxy-D-glucose (95%) is an azide-functionalized glucose analog used as a chemical reporter to visualize and study protein-specific glycosylation in living cells. The azido group at C-6 enables bioorthogonal labeling for downstream detection and imaging in cellular assays.
  • Azide functional group enables bioorthogonal labeling.
  • Suitable for live-cell visualization of protein glycosylation.
  • Supplied as a powder with recommended cold storage for stability.
  • High purity (95%) for consistent assay performance.
  • Defined molecular composition (C6H11N3O5, MW 205.17) for accurate experimental planning.