Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Azido-6-deoxy-D-glucose | 20847-05-6 | MFCD32206193 | 95.0% | 205.17 g/mol | C6H11N3O5 | 5 MG
SDP

Catalog No. 5000284327
Encompass_Preferred

6-Azido-6-deoxy-D-glucose (95%) is an azide-functionalized glucose derivative used as a metabolic probe to label and visualize protein-specific glycosylation in living cells via click chemistry. Supplied as a small-quantity dry powder for research use only.

  • Azide-functionalized glucose probe for metabolic labeling of glycans in living cells.
  • Enables click chemistry-based visualization of protein-specific glycosylation.
  • Provided as a dry powder in small quantities suitable for laboratory experiments.
  • Purity 95% for reliable experimental results.
  • CAS number 20847-05-6; molecular weight 205.17 g/mol.
  • For research use only; not validated for clinical applications.

Catalog No. 50-002-84327 Supplier Medchemexpress LLC Supplier No. HYW6740395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Azido-6-deoxy-D-glucose (95%) is an azide-functionalized glucose derivative used as a metabolic probe to label and visualize protein-specific glycosylation in living cells via click chemistry. Supplied as a small-quantity dry powder for research use only.

  • Azide-functionalized glucose probe for metabolic labeling of glycans in living cells.
  • Enables click chemistry-based visualization of protein-specific glycosylation.
  • Provided as a dry powder in small quantities suitable for laboratory experiments.
  • Purity 95% for reliable experimental results.
  • CAS number 20847-05-6; molecular weight 205.17 g/mol.
  • For research use only; not validated for clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.