Learn More
Medchemexpress LLC 6-azido-hexylamine | 349553-73-7 | 98.7% | 142.20 g/mol | C6H14N4 | 50 MG
SDP
Catalog No. 5000203857
Encompass_Preferred
6-Azido-hexylamine is an azide-functionalized hexylamine used as a cleavable antibody-drug conjugate (ADC) linker and as a click-chemistry reagent for bioconjugation. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) with alkyne-containing molecules and in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO- or BCN-containing partners.
  • Cleavable ADC linker for antibody-drug conjugate synthesis.
  • Azide functional group enables CuAAC and SPAAC bioconjugation.
  • High purity suitable for research and linker-building applications.
  • Available in multiple package sizes, including 50 MG.
Catalog No. 50-002-03857 Supplier Medchemexpress LLC Supplier No. HY13838750MG
6-Azido-hexylamine is an azide-functionalized hexylamine used as a cleavable antibody-drug conjugate (ADC) linker and as a click-chemistry reagent for bioconjugation. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) with alkyne-containing molecules and in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO- or BCN-containing partners.
  • Cleavable ADC linker for antibody-drug conjugate synthesis.
  • Azide functional group enables CuAAC and SPAAC bioconjugation.
  • High purity suitable for research and linker-building applications.
  • Available in multiple package sizes, including 50 MG.