Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-azido-hexylamine | 349553-73-7 | MFCD07369845 | 98.7% | 142.20 g/mol | C6H14N4 | 100 MG
SDP

Catalog No. 5000203933
Encompass_Preferred

6-azido-hexylamine is an azide-functionalized cleavable linker used primarily in antibody-drug conjugate (ADC) synthesis and bioconjugation. The compound contains an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions for attachment to alkyne- or DBCO/BCN-containing molecules.

  • Used as a cleavable ADC linker and bioconjugation reagent.
  • Enables CuAAC and SPAAC click chemistry for efficient coupling.
  • Molecular formula C6H14N4 and molecular weight 142.20 g/mol.
  • High purity (98.7%) and liquid form, colorless to light yellow.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in common laboratory pack sizes including 50 MG and 100 MG.

Catalog No. 50-002-03933 Supplier Medchemexpress LLC Supplier No. HY138387100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-azido-hexylamine is an azide-functionalized cleavable linker used primarily in antibody-drug conjugate (ADC) synthesis and bioconjugation. The compound contains an azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions for attachment to alkyne- or DBCO/BCN-containing molecules.

  • Used as a cleavable ADC linker and bioconjugation reagent.
  • Enables CuAAC and SPAAC click chemistry for efficient coupling.
  • Molecular formula C6H14N4 and molecular weight 142.20 g/mol.
  • High purity (98.7%) and liquid form, colorless to light yellow.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in common laboratory pack sizes including 50 MG and 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.