Learn More
Medchemexpress LLC 6-azido-hexylamine | 349553-73-7 | MFCD07369845 | 98.7% | 142.20 g/mol | C6H14N4 | 250 MG
SDP
Catalog No. 5000204237
Encompass_Preferred
6-Azido-hexylamine is an azide-containing hexylamine used as a cleavable linker and click-chemistry reagent for bioconjugation, including antibody-drug conjugate (ADC) synthesis. It reacts via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), enabling reliable conjugation to alkyne- or cyclooctyne-functionalized partners.
  • Azide functional group for CuAAC and SPAAC reactions.
  • Suitable as a cleavable linker in bioconjugation and ADC workflows.
  • High reported purity for research use.
  • Available in multiple milligram-scale packages for small-scale synthesis.
  • Store protected from light at 4°C; in solvent, long-term storage at -80°C is recommended.
Catalog No. 50-002-04237 Supplier Medchemexpress LLC Supplier No. HY138387250MG
6-Azido-hexylamine is an azide-containing hexylamine used as a cleavable linker and click-chemistry reagent for bioconjugation, including antibody-drug conjugate (ADC) synthesis. It reacts via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), enabling reliable conjugation to alkyne- or cyclooctyne-functionalized partners.
  • Azide functional group for CuAAC and SPAAC reactions.
  • Suitable as a cleavable linker in bioconjugation and ADC workflows.
  • High reported purity for research use.
  • Available in multiple milligram-scale packages for small-scale synthesis.
  • Store protected from light at 4°C; in solvent, long-term storage at -80°C is recommended.