Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-azido-hexylamine | 349553-73-7 | MFCD07369845 | 98.7% | 142.20 g/mol | C6H14N4 | 5 G
SDP

Catalog No. 5000203419
Encompass_Preferred

6-Azido-hexylamine is an azide-containing small molecule used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a click-chemistry reagent. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), and is supplied for research use only.

  • Cleavable ADC linker and click chemistry reagent.
  • Cas number 349553-73-7; molecular formula C6H14N4; molecular weight 142.20 g/mol.
  • Purity 98.7% (manufacturer reported).
  • Physical state: liquid, colorless to light yellow.
  • Available sizes include 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g, and larger quantities by quote.
  • Storage: store at 4°C protected from light; in solution, store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-03419 Supplier Medchemexpress LLC Supplier No. HY1383875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Azido-hexylamine is an azide-containing small molecule used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a click-chemistry reagent. It participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), and is supplied for research use only.

  • Cleavable ADC linker and click chemistry reagent.
  • Cas number 349553-73-7; molecular formula C6H14N4; molecular weight 142.20 g/mol.
  • Purity 98.7% (manufacturer reported).
  • Physical state: liquid, colorless to light yellow.
  • Available sizes include 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g, and larger quantities by quote.
  • Storage: store at 4°C protected from light; in solution, store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.