Learn More
Medchemexpress LLC 6-azidohexan-1-amine | 349553-73-7 | MFCD07369845 | 98.7% | 142.20 g/mol | C6H14N4 | 1 G
SDP
Catalog No. 5000203702
Encompass_Preferred
6-azidohexan-1-amine is an azide-functionalized hexylamine used as a cleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate synthesis. It undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions, and is supplied as a high-purity liquid intended for research use.
  • Azide-functionalized hexylamine suitable for click chemistry.
  • High purity for synthetic and bioconjugation applications.
  • Colorless to light yellow liquid physical form.
  • Stable when stored at 4°C and protected from light; in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • Compatible with copper-catalyzed and strain-promoted click reactions for conjugation to alkynes or DBCO/BCN reagents.
Catalog No. 50-002-03702 Supplier Medchemexpress LLC Supplier No. HY1383871G
6-azidohexan-1-amine is an azide-functionalized hexylamine used as a cleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate synthesis. It undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions, and is supplied as a high-purity liquid intended for research use.
  • Azide-functionalized hexylamine suitable for click chemistry.
  • High purity for synthetic and bioconjugation applications.
  • Colorless to light yellow liquid physical form.
  • Stable when stored at 4°C and protected from light; in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • Compatible with copper-catalyzed and strain-promoted click reactions for conjugation to alkynes or DBCO/BCN reagents.