Learn More
Cayman Chemical 6-AzIdohexnoIc AcId 100mg
SDP
Catalog No. 502967387
Encompass_Preferred
A six carbon saturated fatty acid with an ω-terminal azide group that allows conjugation via click chemistry; commonly used as a linker that is first acylated to one compound to create a derivative that can then be joined to another compound using click chemistry
Catalog No. 50-296-7387 Supplier Cayman Chemical Supplier No. 16518100 MG
A six carbon saturated fatty acid with an ω-terminal azide group that allows conjugation via click chemistry; commonly used as a linker that is first acylated to one compound to create a derivative that can then be joined to another compound using click chemistry