Learn More
Apexbio Technology LLC 6-AZURIDINE-1G
SDP
Catalog No. 5000569000
Encompass_Preferred
6-Azuridine (CAS No 54-25-1) is a nucleoside analog used in biomedical research particularly in antiviral and anticancer studies Originating from modifications of naturally occurring uridine it acts primarily by inhibiting nucleotide biosynthesis interfering with viral RNA replication and targeting pathways involved in cell proliferation Its in vitro activity is typically observed in the nanomolar to low micromolar range making it effective in disrupting viral ion channels and proliferation in cancer cell lines Common applications include use in cell culture models and animal studies to explore its therapeutic potential and to screen for new drugs Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69000 Supplier Apexbio Technology LLC Supplier No. BA43061000
6-Azuridine (CAS No 54-25-1) is a nucleoside analog used in biomedical research particularly in antiviral and anticancer studies Originating from modifications of naturally occurring uridine it acts primarily by inhibiting nucleotide biosynthesis interfering with viral RNA replication and targeting pathways involved in cell proliferation Its in vitro activity is typically observed in the nanomolar to low micromolar range making it effective in disrupting viral ion channels and proliferation in cancer cell lines Common applications include use in cell culture models and animal studies to explore its therapeutic potential and to screen for new drugs Experimental concentrations typically depend on the specific research design