Learn More
Bellco Glass, Inc. 6 Baffle Shake Flask,250mL 38mm Delong Neck
SDP
Catalog No. 501945536
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 250mlDimensions: 80x160mmClosure Sizes: 38Case Quantity: 12
Catalog No. 50-194-5536 Supplier Bellco Glass, Inc. Supplier No. 254360250
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 250mlDimensions: 80x160mmClosure Sizes: 38Case Quantity: 12