Learn More
Bellco Glass, Inc. 6 Baffle Shake Flask, 3000mL 38mm Delong Neck
SDP
Catalog No. 501945540
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 3LDimensions: 182x340mmClosure Sizes: 38Case Quantity: 3
Catalog No. 50-194-5540 Supplier Bellco Glass, Inc. Supplier No. 254363000
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 3LDimensions: 182x340mmClosure Sizes: 38Case Quantity: 3