Learn More
Bellco Glass, Inc. 6 Baffle Shake Flask, 4000mL 38mm Delong Neck
SDP
Catalog No. 501945541
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 4LDimensions: 207x370mmClosure Sizes: 38Case Quantity: 3
Catalog No. 50-194-5541 Supplier Bellco Glass, Inc. Supplier No. 254364000
Manufactured from Type 1 Class A Borosilicate Glass. The 3 side and bottom baffles are used for optimal agitation. 38mm DeLong style neck.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 4LDimensions: 207x370mmClosure Sizes: 38Case Quantity: 3