Learn More
Alkali Scientific 6 BAP [6 Benzylaminopurine], 5 Grams
SDP
Catalog No. 5000661070
Encompass_Preferred
6 Benzylaminopurine (BAP) is a plant growth regulator in powder form, available in a 5g package. BAP is used to promote cell division and shoot formation in plant tissue culture. It is particularly useful for stimulating the growth of adventitious shoots from plant tissues. BAP is commonly used in research and biotechnology to study plant growth and development, as well as in commercial applications to increase plant propagation rates. The powder form allows for easy storage and precise formulation of solutions, ensuring consistent application in experiments and laboratory settings.
Catalog No. 50-006-61070 Supplier Alkali Scientific Supplier No. BAP001CP
6 Benzylaminopurine (BAP) is a plant growth regulator in powder form, available in a 5g package. BAP is used to promote cell division and shoot formation in plant tissue culture. It is particularly useful for stimulating the growth of adventitious shoots from plant tissues. BAP is commonly used in research and biotechnology to study plant growth and development, as well as in commercial applications to increase plant propagation rates. The powder form allows for easy storage and precise formulation of solutions, ensuring consistent application in experiments and laboratory settings.