Learn More
Medchemexpress LLC 6-Benzothiazolamine, N-[6-[2-[(cyclohexylmethyl)amino]-4-(trifluoromethyl)phenyl]-4-pyrimidinyl]- (TFA) | 99.1% | 597.58 | 25 MG
SDP
Catalog No. 5000438226
Encompass_Preferred
AMG 7905 TFA is a hypothermia-inducing transient receptor potential vanilloid type 1 (TRPV1) antagonist. It potentiates TRPV1 channels activation by protons and drives the reflectory inhibition of thermogenesis and tail-skin vasoconstriction, while potently blocking channel activation by capsaicin.
  • Potentiates TRPV1 channels activation by protons
  • Drives reflectory inhibition of thermogenesis and tail-skin vasoconstriction
  • Potently blocks channel activation by capsaicin
Catalog No. 50-004-38226 Supplier Medchemexpress LLC Supplier No. 25MGHY113654A
AMG 7905 TFA is a hypothermia-inducing transient receptor potential vanilloid type 1 (TRPV1) antagonist. It potentiates TRPV1 channels activation by protons and drives the reflectory inhibition of thermogenesis and tail-skin vasoconstriction, while potently blocking channel activation by capsaicin.
  • Potentiates TRPV1 channels activation by protons
  • Drives reflectory inhibition of thermogenesis and tail-skin vasoconstriction
  • Potently blocks channel activation by capsaicin