Learn More
Bioworld 6-Benzylaminopurine (6-BAP), 25 g
Catalog No. 50255037
Encompass_Preferred
6-Benzylaminopurine (6-BAP)Synonyms: N6-Benzyladenine; N-(phenylmethyl)-1H-purin-6-amine Cytokine growth regulatorPlant tissue culture testedSoluble in 1M NaOHSolubilizeat 1 mg/mLin 0.1N KOH
Catalog No. 50-255-037 Supplier Bioworld Supplier No. 306310052
6-Benzylaminopurine (6-BAP)Synonyms: N6-Benzyladenine; N-(phenylmethyl)-1H-purin-6-amine Cytokine growth regulatorPlant tissue culture testedSoluble in 1M NaOHSolubilizeat 1 mg/mLin 0.1N KOH