Learn More
Bioworld 6-Benzylaminopurine Riboside, 250 mg
Catalog No. 50255038
Encompass_Preferred
6-Benzylaminopurine RibosideSynonym: N6-BenzyladenosineCytokine growth regulatorAppearance: white powderMelting Point: 184-186°CSoluble in 1M NaOH
Catalog No. 50-255-038 Supplier Bioworld Supplier No. 306310061
6-Benzylaminopurine RibosideSynonym: N6-BenzyladenosineCytokine growth regulatorAppearance: white powderMelting Point: 184-186°CSoluble in 1M NaOH