Learn More
Bioworld 6-Benzylaminopurine Riboside, 1 g
Catalog No. 50255039
Encompass_Preferred
6-Benzylaminopurine RibosideSynonym: N6-BenzyladenosineCytokine growth regulatorAppearance: white powderMelting Point: 184-186°CSoluble in 1M NaOH
Catalog No. 50-255-039 Supplier Bioworld Supplier No. 306310062
6-Benzylaminopurine RibosideSynonym: N6-BenzyladenosineCytokine growth regulatorAppearance: white powderMelting Point: 184-186°CSoluble in 1M NaOH