Learn More
Abcam 6-Benzylaminopurine, Synthetic cytokinin, 25G
Catalog No. 502704653
Encompass_Preferred
MW 225.26 g/mol. Plant cell division factor. Widely used synthetic cytokinin supplement for plant growth media. Stimulates Ca²+ influx (KM = 1 nM) by moss protoplasts. Induces positive inotropic effects through activation of P2X and P2Y (P2-purinergic receptors) in rat atria. Inhibitor of respiratory kinase in plants, and increases post-harvest life of green vegetables.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4653 Supplier Abcam Supplier No. AB14639825G
MW 225.26 g/mol. Plant cell division factor. Widely used synthetic cytokinin supplement for plant growth media. Stimulates Ca²+ influx (KM = 1 nM) by moss protoplasts. Induces positive inotropic effects through activation of P2X and P2Y (P2-purinergic receptors) in rat atria. Inhibitor of respiratory kinase in plants, and increases post-harvest life of green vegetables.
The product is subject to the following: Abcam Restricted Use Statement