Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-[Bis(2,2,2-trifluoroethyl)amino]-4-(trifluoromethyl)-2(1H)-quinolinone, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013936 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3936 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3936 Supplier MilliporeSigma Supelco Supplier No. 0768225MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 328947-93-9
Linear Formula C14H9F9N2 O
MDL Number MFCD15146764
Melting Point 187°C to 190°C
Synonym LGD-2226
Type Analytical Standard
Quantity 25 mg
Formula Weight 392.22
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.