Learn More
Apexbio Technology LLC 6-Bnz-cAMP sodium salt(Synonyms: N6-Benzoyl-cAMP sodium salt, 6-Benzoyladenosine-3',5'-cyclic monophosphate sodium salt), 10mg, CAS: 1135306-29-4.
SDP
Catalog No. 5000061227
Encompass_Preferred
6-Bnz-cAMP sodium salt (CAS 1135306-29-4) is a selective cAMP analogue that functions as an activator of protein kinase A (PKA)-dependent signaling pathways It specifically targets the conserved cAMP-binding domains on PKA triggering downstream phosphorylation events Activation of PKA subsequently impacts signaling proteins such as CREB whose phosphorylation state mediates processes like osteoblast differentiation Experimental studies indicate 6-Bnz-cAMP promotes differentiation mineralization and early cell adhesion notably it drives osteogenic differentiation in MC3T3-E1 cells Its potential for controlled release further suggests utility as a molecule for regenerative strategies in bone tissue engineering
Catalog No. 50-000-61227 Supplier Apexbio Technology LLC Supplier No. B779110
6-Bnz-cAMP sodium salt (CAS 1135306-29-4) is a selective cAMP analogue that functions as an activator of protein kinase A (PKA)-dependent signaling pathways It specifically targets the conserved cAMP-binding domains on PKA triggering downstream phosphorylation events Activation of PKA subsequently impacts signaling proteins such as CREB whose phosphorylation state mediates processes like osteoblast differentiation Experimental studies indicate 6-Bnz-cAMP promotes differentiation mineralization and early cell adhesion notably it drives osteogenic differentiation in MC3T3-E1 cells Its potential for controlled release further suggests utility as a molecule for regenerative strategies in bone tissue engineering