Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Boc-1-oxa-6-azaspiro[3.3]heptane, 96%, Thermo Scientific Chemicals

Catalog No. AAH57040MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57040MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57040MD Supplier Thermo Scientific Chemicals Supplier No. H57040MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1223573-41-8 | Molecular Formula: C10H17NO3 | Molecular Weight: 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1223573-41-8
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD16877674
InChI Key QOXFRPSIASGKGE-UHFFFAOYSA-N
Synonym tert-butyl 1-oxa-6-azaspiro 3.3 heptane-6-carboxylate, 1-oxa-6-azaspiro 3.3 heptane-6-carboxylic acid tert-butyl ester, 6-boc-1-oxa-6-azaspiro 3.3 heptane, tert-butyl2,5-dioxa-8-aza-spiro 3,5 nonane-8-carboxylate, 2-aza-5-oxaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester, 1-oxa-6-azaspiro 3.3 heptane-6-carboxylic acid,1,1-dimethylethyl ester, tert-butyl 1-oxa-6-azaspiro 3.3 heptane-6-carboxylate 95+%
PubChem CID 54758968
IUPAC Name tert-butyl 1-oxa-6-azaspiro[3.3]heptane-6-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2(CCO2)C1

Specifications

Quantity 250 mg
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material 6-Boc-1-oxa-6-azaspiro[3.3]heptane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.