Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Boc-2-thia-6-aza-spiro[3.3]heptane-2,2-dioxide

Catalog No. PHB00020 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PHB00020 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PHB00020 Supplier Sigma Aldrich Supplier No. PHB00020100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(benzyloxy)ethanol has been used in:
  • 9-(2-hydroxyethyl)-linked synthesis of 2,6-diaminopurines
  • Synthesis of benzyloxy ethyl methacrylate monomer by esterification with methacryloyl chloride
  • A base-free iridium-catalyzed direct alkylation of active methylene compounds with alcohols

Specifications

CAS 1291487-31-4
MDL Number MFCD18839239
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.