Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-1,8-naphthyridin-2(1H)-one, 98%

Catalog No. AAH50042MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50042MD 250 mg
AAH5004203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50042MD Supplier Thermo Scientific Chemicals Supplier No. H50042MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 72754-05-3 | C8H5BrN2O | 225.05 g/mol

6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1,8-naphthyridin-2(1H)-one is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72754-05-3
Molecular Formula C8H5BrN2O
Molecular Weight (g/mol) 225.05
MDL Number MFCD07781151
InChI Key BCEWGGGNOQNYKK-UHFFFAOYSA-N
PubChem CID 12520144
IUPAC Name 6-bromo-1H-1,8-naphthyridin-2-one
SMILES BrC1=CN=C2NC(=O)C=CC2=C1

Specifications

Melting Point >300°C
Quantity 250 mg
Synonym 6-bromo-1,8-naphthyridin-2-ol, 6-bromo-1,8-naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2 1h-one, 6-bromo-1,8 naphthyridin-2-ol, 6-bromo-2-hydroxy-1,8-naphthyridine, 1,8-naphthyridin-2 1h-one, 6-bromo, acmc-1bihh, ksc496s1t, 6-bromopyridino 2,3-b pyridin-2-ol, 6-bromo-1,8naphthyridin-2 1h-one
Solubility Information Insoluble in water.
Formula Weight 225.05
Percent Purity 98%
Chemical Name or Material 6-Bromo-1,8-naphthyridin-2(1H)-one
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.