Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Bromo-1,8-naphthyridin-2-amine

Catalog No. CDS019239 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS019239 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS019239 Supplier Sigma Aldrich Supplier No. CDS019239100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-aminotoluene-3-sulfonic acid acts as schiff base ligand and forms binary and ternary complexes with co(ii), ni(ii), cu(ii) and zn(ii) ions.4-aminotoluene-3-sulfonic acid was used to investigate the enrichment of aminobenzoic acids and aminotoluenesulfonic acids using graphitized carbon black (gcb) by solid-phase extraction
  • It was also used in the synthesis of azobenzene derivatives.dupont product

Specifications

Molecular Formula C8H6BrN3
Linear Formula C8H6BrN3
MDL Number MFCD09909648
Quantity 250 g
Molecular Weight (g/mol) 224.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.