Learn More
Medchemexpress LLC 6-Bromo-1-hexanol | 4286-55-9 | 500g
SDP
Catalog No. 5000301793
Encompass_Preferred
6-Bromo-1-hexanol posesses a terminal bromine and hydroxylic group The bromine is a very good leaving group for nucleophilic substitution reactions The hydroxyl group enables further derivatization or replacement with other reactive functional groups
Catalog No. 50-003-01793 Supplier Medchemexpress LLC Supplier No. HYW016455500G
6-Bromo-1-hexanol posesses a terminal bromine and hydroxylic group The bromine is a very good leaving group for nucleophilic substitution reactions The hydroxyl group enables further derivatization or replacement with other reactive functional groups