Learn More
Medchemexpress LLC 6-bromo-1-hexanol | 4286-55-9 | MFCD00002983 | 96.7% | 181.07 g/mol | C6H13BrO | 25 G
SDP
Catalog No. 5000304099
Encompass_Preferred
6-Bromo-1-hexanol is a halogenated primary alcohol used as a synthetic intermediate and biochemical reagent. It contains a terminal bromine and a primary hydroxyl group, enabling nucleophilic substitution and further functionalization for use in organic synthesis and analytical derivatization. Batch-specific documentation is provided for purity verification.
  • Terminal bromine enables nucleophilic substitution reactions.
  • Primary hydroxyl group supports derivatization and functional group transformations.
  • Useful building block for organic synthesis and analytical derivatization workflows.
  • Colorless to light-yellow liquid with molecular formula C6H13BrO and molecular weight 181.07 g/mol.
  • Batch-specific certificate of analysis available for purity verification.
Catalog No. 50-003-04099 Supplier Medchemexpress LLC Supplier No. 25GHYW016455
6-Bromo-1-hexanol is a halogenated primary alcohol used as a synthetic intermediate and biochemical reagent. It contains a terminal bromine and a primary hydroxyl group, enabling nucleophilic substitution and further functionalization for use in organic synthesis and analytical derivatization. Batch-specific documentation is provided for purity verification.
  • Terminal bromine enables nucleophilic substitution reactions.
  • Primary hydroxyl group supports derivatization and functional group transformations.
  • Useful building block for organic synthesis and analytical derivatization workflows.
  • Colorless to light-yellow liquid with molecular formula C6H13BrO and molecular weight 181.07 g/mol.
  • Batch-specific certificate of analysis available for purity verification.