Learn More
Medchemexpress LLC 6-bromo-1-hexanol | 4286-55-9 | MFCD00002983 | 98.5% | 181.07 | C6H13BrO | 1 ML
SDP
Catalog No. 5000303767
Encompass_Preferred
6-bromo-1-hexanol is an organobromide with a terminal hydroxyl group used as a synthetic building block in organic and polymer chemistry and as a biochemical reagent. The bromide functions as a strong leaving group for nucleophilic substitution and cross-linking reactions. It is supplied as bulk liquid and as a 10 mM solution in DMSO for ready use in derivatization and intermediate synthesis.
  • Terminal bromide and hydroxyl functional groups enable versatile transformations
  • Suitable for nucleophilic substitution and cross-linking reactions
  • Available as bulk liquid and ready-to-use solution formats
  • Documented purity with certificate of analysis available
  • Storage recommendations provided for pure and in-solution material
Catalog No. 50-003-03767 Supplier Medchemexpress LLC Supplier No. HYW01645510MM/1ML
6-bromo-1-hexanol is an organobromide with a terminal hydroxyl group used as a synthetic building block in organic and polymer chemistry and as a biochemical reagent. The bromide functions as a strong leaving group for nucleophilic substitution and cross-linking reactions. It is supplied as bulk liquid and as a 10 mM solution in DMSO for ready use in derivatization and intermediate synthesis.
  • Terminal bromide and hydroxyl functional groups enable versatile transformations
  • Suitable for nucleophilic substitution and cross-linking reactions
  • Available as bulk liquid and ready-to-use solution formats
  • Documented purity with certificate of analysis available
  • Storage recommendations provided for pure and in-solution material