Learn More
Medchemexpress LLC 6-bromo-1-hexanol | 4286-55-9 | MFCD00002983 | 98.5% | 181.07 g/mol | C6H13BrO | 100 G
SDP
Catalog No. 5000303322
Encompass_Preferred
6-Bromo-1-hexanol is a primary bromoalcohol used as an intermediate in organic synthesis and small-molecule assembly. It features a terminal bromine that serves as an effective leaving group and a primary hydroxyl that enables a wide range of functional-group transformations.
  • Useful in nucleophilic substitution reactions due to terminal bromine.
  • Hydroxyl group enables derivatization, esterification, and etherification.
  • Liquid form facilitates handling and reagent addition.
  • Available in multiple package sizes to support discovery and scale-up.
  • Molecular formula C6H13BrO; molecular weight 181.07 g/mol.
Catalog No. 50-003-03322 Supplier Medchemexpress LLC Supplier No. HYW016455100G
6-Bromo-1-hexanol is a primary bromoalcohol used as an intermediate in organic synthesis and small-molecule assembly. It features a terminal bromine that serves as an effective leaving group and a primary hydroxyl that enables a wide range of functional-group transformations.
  • Useful in nucleophilic substitution reactions due to terminal bromine.
  • Hydroxyl group enables derivatization, esterification, and etherification.
  • Liquid form facilitates handling and reagent addition.
  • Available in multiple package sizes to support discovery and scale-up.
  • Molecular formula C6H13BrO; molecular weight 181.07 g/mol.