Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Bromo-1-hexene

Catalog No. 2472195G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-721-95G 5 g
24-721-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-721-95G Supplier Sigma Aldrich Supplier No. 2472195G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reaction of 6-bromo-1-hexene with mg to form grignard compounds has been investigated
  • The thermal chemistry of 6-bromo-1-hexene on ni (100) surface has been studied under ultrahigh vacuum conditions by x-ray photoelectron spectroscopy and temperature-programmed desorption.6-bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane
  • It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia

Specifications

CAS 2695-47-8
Density 1.22 g/mL (at 25°C (literature))
Boiling Point 47°C to 51°C (16 mmHg)
Molecular Formula C6H11Br
Refractive Index n20/D 1.465 (literature)
Linear Formula Br(CH2)4 CH=CH2
MDL Number MFCD00000269
Quantity 5 g
Molecular Weight (g/mol) 163.06
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.