Learn More
Sigma Aldrich 6-Bromo-2,2-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Reactant for:
- Two-step syntheses of fused quinoxaline-benzodiazepines and bis-benzodiazepines employing a double udc (ugi/deprotect/cyclize) strategy
- Preparation of hydroquinazoline scaffolds via microwave-promoted ugi reaction and cyclization
- Preparation of pyrazinone inhibitors of mast cell tryptase
Specifications
Specifications
| Molecular Formula | C10H10BrNO2 |
| Linear Formula | C10H10BrNO2 |
| MDL Number | MFCD12196527 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 256.1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.