Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2,3-dicyanonaphthalene 98.0+%, TCI America™
SDP

Catalog No. B13901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13901G Supplier TCI America Supplier No. B13901G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70484-02-5
Molecular Formula C12H5BrN2
Molecular Weight (g/mol) 257.09
MDL Number MFCD00130113
InChI Key UERLNOIJNRUJKQ-UHFFFAOYSA-N
Synonym 6-Bromonaphthalene-2,3-dicarbonitrile
PubChem CID 784710
IUPAC Name 6-bromonaphthalene-2,3-dicarbonitrile
SMILES BrC1=CC=C2C=C(C#N)C(=CC2=C1)C#N

Specifications

Melting Point 252°C
Quantity 1 g
UN Number 3439
Formula Weight 257.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2,3-dicyanonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.