Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-benzothiazolinone 97.0+%, TCI America™
SDP

Catalog No. B20121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20121G Supplier TCI America Supplier No. B20121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62266-82-4
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.08
MDL Number MFCD00239363
InChI Key HECJMTPEVWQFCY-UHFFFAOYSA-N
Synonym 6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
PubChem CID 188444
IUPAC Name 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one
SMILES BrC1=CC=C2NC(=O)SC2=C1

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 1 g
Formula Weight 230.08
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-benzothiazolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.