Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Bromo-2-(bromomethyl)-3-fluoropyridine | 1187836-89-0 | 96.79% | 268.91 | 50 MG
SDP

Catalog No. 5000405204
Encompass_Preferred

6-Bromo-2-(bromomethyl)-3-fluoropyridine is a biological molecule intended for research use only. It is not for human use or sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-05204 Supplier Medchemexpress LLC Supplier No. HY4372750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Bromo-2-(bromomethyl)-3-fluoropyridine is a biological molecule intended for research use only. It is not for human use or sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.